missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One F-Bottom 96-well Assay Microplates
Features a chimney-like arrangement of the wells that allows each well to stand on its own. Greiner Bio One™ F-Bottom 96-Well Assay Microplates are constructed of polystyrene, a highly clear polymer with excellent optical properties making it ideal for precise optical measurements. Polystyrene is also characterized by its ability to bind biomolecules, such as proteins and is suitable for work with cell cultures.
156.00 € - 321.00 €
Specifications
Certifications/Compliance | DIN ISO 9001 |
---|---|
Certified | RNase-/D-Nase, human DNA free |
Format | Standard |
Culture Area | 34 mm2 |
Array | 8 x 12 |
Product Code | Brand | Color | Sterility | Lid | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Sterility | Lid | Price | Quantity & Availability | |||
10284272
|
Greiner Bio-One
655083 |
White | Sterile | With Lid |
321.00 €
Pack of 32 |
In Stock
Log in to see stock. |
|||
10164911
|
Greiner Bio-One
655074 |
White | Sterile | Without Lid |
156.00 €
Pack of 40 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable endotoxins (0.003 EU/mL) and regularly tested using an FDA-approved kinetic turbidimetric LAL-test (Limulus Amoebocyte Assay)
- Manufactured from raw materials tested for leachables
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
Specifications
DIN ISO 9001 | |
Standard | |
8 x 12 | |
96 | |
392 μL | |
Yes | |
11.5 mm |
RNase-/D-Nase, human DNA free | |
34 mm2 | |
Polystyrene | |
25 to 34 μL | |
Flat Bottom (F-well) | |
Yes | |
5 mm |
Safety and Handling
missing translation for 'shelfLife' : 4 Years after month of production