Learn More
Julabo™ F Series Recirculating Cooling Baths with Air Compressor
Require very little laboratory space and have very low procurement costs. . Julabo™ F Series Recirculating Cooling Baths with Air Compressor are a great way to replace costly systems based on running tap water and are ideal for basic cooling tasks.
Brand:  Julabo™ 9620025
 View more versions of this product
Product Code. 11994858
  • 3230.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Environmentally friendly operation with low energy consumption
  • Compact design
  • Splashproof membrane keypad with LED temperature display
  • Straightforward filling and draining
  • Filling level indicator
  • May be used with water, water/glycol, Julabo Thermal G
Specifications
Specifications
2.6 L
0.09kW at -5°C, 0.18kW at 0°C, 0.21kW at 5°C, 0.22kW at 10°C, 0.25kW at 20°C (Me
-10°C to +40°C
F250
±0.5°C
40 cm
52 cm
27 kg
115/230 V
UL Listed and NRTL marked
15 L/min.
50.7 psi
R134A
±0.5°C
24 cm
27.66 kg
115/230 V 60 Hz 6 A
60 Hz