missing translation for 'onlineSavingsMsg'
Learn More

Julabo™ F Series Recirculating Cooling Baths with Air Compressor

Require very little laboratory space and have very low procurement costs. . Julabo™ F Series Recirculating Cooling Baths with Air Compressor are a great way to replace costly systems based on running tap water and are ideal for basic cooling tasks.

Brand:  Julabo™ 9620050

4197.58 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15194342

  • 4605.00 € / Each
Estimated Shipment: 08-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Environmentally friendly operation with low energy consumption
  • Compact design
  • Splashproof membrane keypad with LED temperature display
  • Straightforward filling and draining
  • Filling level indicator
  • May be used with water, water/glycol, Julabo Thermal G
Specifications

Specifications

7.5 L
0.25kW at 0°C, 0.3kW at 5°C, 0.4kW at 10°C, 0.5kW at 20°C (Medium Ethanol)
0°C to 40°C
F500
±0.5°C
44 cm
59 cm
34 kg
115/230 V
CE
24 L/min.
7.25 psi
R134A
±0.5°C
37.5 cm
49.44 kg
115/230 V 60 Hz
60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers