missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MULTIROIR F64 Trolley
Red trolley F64 for plastic bin with 4 swivel wheels
Brand: MULTIROIR ARO64RGU
This item is not returnable.
View return policy
Description
- Red trolley F64
- For plastic bin 600 x 400
- Material: ABS
- Load capacity: 250 kg
- Mobility: 4 swivel wheels
- External dimensions: 620 x 420 mm
Specifications
250 kg | |
620 x 420 mm | |
Transport All Types of Products | |
600 x 400 Euro-Standard Container |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction