missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Fabric Coveralls With Hood
BIOPHARMA

Product Code. 12797186
Click to view available options
Size:
Small
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

The BioClean-D™ coverall with hood is a disposable garment featuring a front zip with protective flap, elasticated hood, back, cuffs and ankles and thumb loops to ensure a secure hold. The BioClean-D coverall provides exceptional comfort and protection, even when handling cytotoxic drugs

  • Low-linting & durable CleanTough™material
  • Thumb loops to ensure a secure hold
  • Zip with sealable cover
  • Elasticated hood, back, cuffs and ankles
  • Tested against permeation standard ASTM D6978-05
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Medium
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper
Sterility Non-sterile
Resistance Isopropyl Alcohol
Back Style Elastic
Certifications/Compliance ISO 9001:2008
Description Zip front with flap cover, elasticated hood, back, cuffs and ankles
Size (Metric) Chest 914.4 to 990.6 mm
For Use With (Application) Controlled Environments
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'