missing translation for 'onlineSavingsMsg'
Learn More

Moldex™ AIR FFP2 Face Masks

Provides protection against dust, mist, fume.

Brand:  Moldex™ 310501

Product Code. 12962415

  • 118.00 € / Pack of 10
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • FFP2 Mask with pleated filter technology: 260% more filter surface area
  • Significantly reduced breathing resistance
  • High filter capacity, longer usage time
  • Available in two sizes
  • DuraMesh™: strong and durable structure, keeps mask in shape
  • ActivForm™: no nose clip needed
  • Comfortable nose seal
  • Loop strap with clip for quick, simple and safe on and off
  • Tested and certified to EN 149:2001+A1:2009
  • Protection level: FFP2 NR D
  • FFP2: 10 x Workplace Exposure Limit
  • NR [non reusable]: Disposable FFP. Comfortable and durable throughout the whole shift.
  • D: These masks have passed the optional Dolomite clogging test. Better breathing resistance for longer.
Specifications

Specifications

General Purpose Face Mask
Medium/Large
DuraMesh™: Strong and Durable Structure, Keeps the Mask in Shape
Single-use
Respiratory Protection, Prevent Particles (droplets)
Airborne Contaminants, Reduced Breathing Resistance
White
EN 149:2001 + A1:2009
AIR FFP Face Mask, Respiratory Protection, Single use only
2.6
Loop strap, High filter, Ventex valve
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers