Learn More
Moldex™ Face Masks
Click to view available options
Size:
Medium/Large
Small
Unit Size:
Pack of 10
Description
The uniquely developed Series 3000 Air from Moldex offers extremely low breathing resistance and therefore increased performance of both mask and wearer. The filtering surface is almost three times larger than a standard filter product, allowing you to breathe more easily.
Specifications
Specifications
| Color | White |
| Product Type | Face Mask |
| Size | Small |
| Quantity | 10/Pk. |