missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Factor XIIIa Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17223-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the coagulation factor XIII A subunit. Coagulation factor XIII is the last zymogen to become activated in the blood coagulation cascade. Plasma factor XIII is a heterotetramer composed of 2 A subunits and 2 B subunits. The A subunits have catalytic function, and the B subunits do not have enzymatic activity and may serve as plasma carrier molecules. Platelet factor XIII is comprised only of 2 A subunits, which are identical to those of plasma origin. Upon cleavage of the activation peptide by thrombin and in the presence of calcium ion, the plasma factor XIII dissociates its B subunits and yields the same active enzyme, factor XIIIa, as platelet factor XIII. This enzyme acts as a transglutaminase to catalyze the formation of gamma-glutamyl-epsilon-lysine crosslinking between fibrin molecules, thus stabilizing the fibrin clot. It also crosslinks alpha-2-plasmin inhibitor, or fibronectin, to the alpha chains of fibrin. Factor XIII deficiency is classified into two categories: type I deficiency, characterized by the lack of both the A and B subunits; and type II deficiency, characterized by the lack of the A subunit alone. These defects can result in a lifelong bleeding tendency, defective wound healing, and habitual abortion.Specifications
Factor XIIIa | |
Polyclonal | |
Unconjugated | |
F13A1 | |
Coagulation factor XIIIa, F13A, F13A1, Transglutaminase A chain | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2162, 60327, 74145 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08619, P00488, Q8BH61 | |
F13a1 | |
Factor XIIIa Fusion Protein Ag11059 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |