Learn More
Memmert™ Factory Calibration (3 Temperatures) for UF30plus
Calibration at Memmert factory for three temperature value.
Brand:  Memmert™ UF30plusD00128
Product Code. 17599275
  • 356.00 € / Each
Explore more special offers
Description
Description
  • For three temperature values: 100°C, 160°C, and 220°C
Specifications
Specifications
UF30plus Oven
Measuring point chamber center
Certificate for 3 temperatures
Standard Works Calibration Certificate