missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Bacteriological Petri Dishes with Lid
Manufactured from crystal-grade polystyrene for optical clarity. Falcon™ Bacteriological Petri Dishes are not tissue culture treated for cell attachment for applications where cell attachment is not desired. The surface is hydrophobic.
Brand: Falcon™ 351029
This item is not returnable.
View return policy
Description
These bacteriological Petri dishes are not tissue culture treated for cell attachment; for applications where cell attachment is not desired. Made from crystal-grade polystyrene for optical clarity. Surface is hydrophobic.
- Durable construction for stable dish manipulation
- Crystal-grade, virgin polystyrene for optical clarity
- Sterile
- Dimensions listed (diameter x height)
Specifications
56.5 cm2 | |
Polystyrene | |
Non-treated | |
500/Cs. | |
15 mm |
Single-use | |
Sterile | |
Cell Culture Petri Dish | |
20/Pk. | |
100 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction