missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Cell Culture Inserts
BIOPHARMAGREENER_CHOICE

Product Code. 10136410 Shop All Corning Products
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
Color:
Translucent
Transparent
Unit Size:
Pack of 48
This item is not returnable. View return policy

Product Code. 10136410

Brand: Falcon™ undefined

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Use with 24-well plates. Falcon™ Cell Culture Inserts (cell culture on permeable supports) results in greater morphological and/or physiological differentiation of many cell types.

  • Track-etched PET membranes have a smooth surface and defined cylindrical pores that traverse the membrane
  • Low protein binding PET membrane
  • Sterilized by gamma irradiation
  • A wide variety of configurations including 6, 12, and 24 well
  • A broad selection of membrane pore sizes, 0.4, 1.0, 3.0, and 8.0 μm diameter
  • Packed in individual blister packs, 48 inserts/case
  • Non-Tissue Culture-treated insert housings prevent promiscuous growth of cells on the insert walls
  • Innovative hanging design facilitates pipetting and allows for co-culture

Order Info

Individually packaged in color-coded, peel-open medical-style packaging

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 0.3 cm2
Material PET
Pore Density 6±2 x 104 pores/cm2
Pore Size 8 μm
Type Membrane
For Use With (Equipment) 24 Well Plate
Diameter (Metric) Filter 6.4 mm
No. per Pack 8/Pk.
No. per Case 48/Cs.
Surface Coating TC-Treated
Color Transparent
Surface Treatment TC-Treated
Sterility Sterile
No. of Wells 24
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.