Learn More
Falcon™ Cell Culture Multi Flasks
BIOPHARMAGREENER_CHOICE
Grow cells faster and easier, increasing productivity in cell culture workflow. Falcon™ Cell Culture Mini Flasks provide even distribution of media across all layers for homogeneous cell growth.
Brand:  Falcon™ 353143
 View more versions of this product
Product Code. 11587421
  • 164.00 € / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.
  • Even distribution of media across all layers for homogeneous cell growth
  • Ability to mix cells and reagents in the Falcon Multi-Flask saves time
  • Flexible design allows to pour or aspirate/recover cells using a pipet
  • Consistent surface treatment for predictable scale-up
  • Lot number printed on each flask for traceability
  • Manufactured in compliance to cGMP standards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polystyrene
Rectangular
Sterile
525 cm2
Straight
3 Layer TC-Treated
Vented
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.