Learn More
Falcon™ Polystyrene Aspirating Pipets, Without Graduations
BIOPHARMAGREENER_CHOICE
Manufactured from polystyrene as a safer alternative to glass Pasteur pipets. Falcon™ Disposable Aspirating Pipets are ideal for tissue culture usage or any non-toxic solution that requires aliquoted or aspirating/dispensing in a sterile environment.
Brand:  Falcon™ 357501
 View more versions of this product
Product Code. 10635611
  • 64.98 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Sterile, polystyrene, non-plugged, non-graduated pipets
  • Safer alternative to glass Pasteur pipets
  • Can be used for all vacuum-aspirating procedures
  • Noncytotoxic, nonpyrogenic tested to less than 0.1 EU/mL
  • Individually packaged in thermoformed paper-plastic wrap to ensure sterile presentation
Order Info
All sizes come individually wrapped, 200 per case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Class I Medical
Polystyrene
For tissue culture usage or any non-toxic solution that requires aliquot or aspirating, dispensing in a sterile environment
5 mL
Single-use
Aspirating
Sterile
13.71 in.
Aspirating Pipet