missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM49B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20127-1-AP-150UL
This item is not returnable.
View return policy
Specifications
FAM49B | |
Polyclonal | |
Unconjugated | |
FAM49B | |
BM 009, FAM49B, L1, Protein FAM49B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
223601, 299909, 51571 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q921M7, Q9NUQ9 | |
FAM49B | |
FAM49B Fusion Protein Ag13939 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction