Learn More
FAM49B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20127-1-AP-20UL
 View more versions of this product
Product Code. 16839624
  • 145.00 € / 20µL
Explore more special offers
Specifications
FAM49B
Polyclonal
Unconjugated
FAM49B
BM 009, FAM49B, L1, Protein FAM49B
Rabbit
Antigen Affinity Chromatography
RUO
223601, 299909, 51571
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q921M7, Q9NUQ9
FAM49B
FAM49B Fusion Protein Ag13939
20 μL
Primary
Mouse, Human, Rat