missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Blue BB, 98%, pure

101.51 € - 302.51 €

Chemical Identifiers

CAS 120-00-3
Molecular Formula C17H20N2O3
Molecular Weight (g/mol) 300.358
MDL Number MFCD00009091
InChI Key CNXZLZNEIYFZGU-UHFFFAOYSA-N
Synonym 4'-Amino-2', 5'-diethoxybenzanilide, Azoic Diazo No.20, C.I. 37175
PubChem CID 67108
IUPAC Name N-(4-amino-2,5-diethoxyphenyl)benzamide
SMILES CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10052610
View Documents Promotion Details
Thermo Scientific Chemicals
191570250
25 g Glass bottle
101.51 €
25g
In Stock
to see stock.
Add to basket
 
10216490
View Documents Promotion Details
Thermo Scientific Chemicals
191571000
100 g Glass bottle
302.51 €
100g
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

120-00-3
300.358
CNXZLZNEIYFZGU-UHFFFAOYSA-N
67108
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
C17H20N2O3
MFCD00009091
4'-Amino-2', 5'-diethoxybenzanilide, Azoic Diazo No.20, C.I. 37175
N-(4-amino-2,5-diethoxyphenyl)benzamide

Specifications

98.0°C to 100.0°C
98%
MFCD00009091
4'-Amino-2', 5'-diethoxybenzanilide, Azoic Diazo No.20, C.I. 37175
Authentic
N-(4-amino-2,5-diethoxyphenyl)benzamide
67108
98%
Complies
Powder or Chunks
120-00-3
C17H20N2O3
13, III, 2170
CNXZLZNEIYFZGU-UHFFFAOYSA-N
CCOC1=CC(=C(C=C1N)OCC)NC(=O)C2=CC=CC=C2
300.358
300.35
Pure
Brown to Gray
Fast blue BB, 98%
Videos
Safety and Handling

Safety and Handling

GHS P Statement
Fatal if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

missing translation for 'einecsNumber' : 204-363-8

missing translation for 'tsca' : TSCA

Product Identifier
  • Fast Blue BB

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only