Learn More
Epredia™ Fast Flex™ Holding Solution
Product Code. p-4532395 Shop All Epredia Products
Click to view available options
Quantity:
5L
2.5 gal.
Unit Size:
5 L
Each
Product Code. 12559946
Brand: Epredia™ 9800
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Employ a fixative designed for use in the grossing room with Fast Flex™ Holding Solution.
  • Place cassettes in holding solution for at least 20 minutes prior to loading on the processor
  • Compatible with 10% Neutral Buffered Formalin and other fixatives
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2.5 gal.