missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Fast-Flex™ Processing Reagents
Improve the quality of processed tissue with these processing reagents, which can either be ordered with a xylene-based or a xylene-substitute-based clearant.
Brand: Epredia™ 9802
This item is not returnable.
View return policy
Description
- Reduces Tissue processing time by 80%
- Decreases the amount of reagents used up to 75%
- Simplifies reagent management by eliminating the need to rotate reagents
- Compatible with H&E, IHC, Special Stains and In situ
Includes:
One 5L bottle each: Fixative (Solution 1), Dehydrant (Solution 2) and Clearing Reagent (Solution 3).

Specifications
Xylene-free Clearant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Fast-Flex™ Processing Reagents > Xylene-free clearant
Spot an opportunity for improvement?Share a Content Correction