missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastAP Thermosensitive Alkaline Phosphatase (1 U/μL)

Catalyze the removal of 5'- and 3'-phosphate groups from DNA, RNA, nucleotides, and proteins using this thermosensitive alkaline phosphatase .

43.42 € - 362.00 €

Specifications

Concentration 1 U/μL
Enzyme Phosphatase
Compatible Buffer PCR Buffer, Enzyme Buffer
Product Type Alkaline Phosphatase
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10151000
View Documents
Thermo Scientific™
EF0651
1,000 units
97.00 €
1000 units
Please to purchase this item. Need a web account? Register with us today!
10699730
View Documents
Thermo Scientific™
EF0652
5 x 1,000 units
362.00 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
10819230
View Documents
Thermo Scientific™
EF0654
300 units
43.42 €
300 units
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FastAP Thermosensitive Alkaline Phosphatase catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA, and nucleotides. This enzyme also removes phosphate groups from proteins.

FastAP is a novel alkaline phosphatase, which is active in all Thermo Scientific restriction enzyme buffers as well as in PCR buffers. It dephosphorylates all types of DNA ends (blunt, 5'- and 3'-overhangs) in 10 minutes at 37°C. The enzyme is inactivated in 5 minutes at 75°C . Therefore, removal of alkaline phosphatase is not required prior to ligation.

Highlights

  • Recombinant enzyme
  • Fast dephosphorylation -10 minutes at 37°C
  • Fast and complete inactivation -5 minutes at 75°C
  • Simultaneous digestion and dephosphorylation of vector DNA
  • 100% active in restriction enzyme and PCR buffers
  • One protocol for all types of DNA ends:
    • 5'-overhangs
    • 3'-overhangs
    • blunt-ends
    • single nucleotides
  • PCR clean-up in conjunction with Exo I
  • Protein dephosphorylation
Specifications

Specifications

1 U/μL
Phosphatase
PCR Buffer, Enzyme Buffer
Alkaline Phosphatase
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.