missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest Green Buffer (10X)

Proprietary digestion buffer which supports 100% activity of all FastDigest restriction enzymes.

Brand:  Thermo Scientific™ B72

16.47 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 11836863

  • 20.85 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

DNA/RNA modifying enzymes, such as ligases, phosphatases, kinases and mesophilic DNA polymerases also have 100% activity in FastDigest Green Buffer. Therefore, enzymes used in downstream applications can be directly added to the FastDigest reaction mix.

  • Double and multiple digestion in a single buffer with any combination of enzymes
  • No sequential digestions or buffer changes needed
  • Allows direct loading of reaction mixture on gels
  • Saves pipetting time and eliminates the need for DNA loading dyes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10X
Digestion Buffer
FastDigest™
5 x 1 mL
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2877012 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberB72
Lot Number2877082 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberB72
Lot Number3143233 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog NumberB72
Lot Number2780334 Certificate TypeCertificate of Analysis Date08/08/2023 Catalog NumberB72
Lot Number2704412 Certificate TypeCertificate of Analysis Date14/04/2023 Catalog NumberB72
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FastDigest Green Buffer (10X) >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.