Learn More
Thermo Scientific™ FastDigest™ NheI: PROMO
Thermo Scientific FastDigest NheI restriction enzyme recognizes G^CTAGC site and cuts best at 37°C in 5 to 15 minutes using universal FastDigest Buffer. Isoschizomers: AsuNHI, BmtI.
Brand:  Thermo Scientific™ FD0973
Product Code. 10755851
  • 46.97 € / 50 reactions
Explore more special offers
Description
Description
  • 100% activity of all FastDigest enzymes in the universal buffer
  • 100% buffer compatibility with downstream applications
  • 100% buffer compatibility with downstream applications
  • Direct loading on gels
  • Direct loading on gels
  • 176 FastDigest enzymes available
TRUSTED_SUSTAINABILITY
Specifications
Specifications
37°C
G^CTAGC
Restriction Enzyme
FastDigest™