Learn More
MP Biomedicals™ FastGro Synthetic, Animal Free, Chemically Defined FBS Replacement
FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound.
Brand: MP Biomedicals™ 092640054
1299.69 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
FastGro allows culturing cells in vitro without the use of serum and without any animal or human derived compound. FastGro is a fully defined additive, avoiding the need for serum, for a wide range of cell culture practices. Since FastGro is chemically defined it avoids lot-to-lot variations. All components in FastGro are highly purified and identified chemical compounds.
Culturing cells in a FastGro enriched medium represents a number of benefits as compared to conventional FBS enriched media.
- Due to its chemically defined nature there are no lot-to-lot variations. This skips the need for testing new batches of serum.
- There is no interference of unknown serum compounds in your research.
- Since it doesn't contain hormones nor cytokines it doesn't bias your research data.
- FastGro is save in use, since it doesn't contain animal or human derived materials.
- It can be stored in the refrigerator. No need for thawing before use.
Specifications
Store at 2 to 8°C | |
Synthetic sera | |
500 mL | |
Cell Biology | |
Cell Culture |
Liquid | |
FBS Replacement Serum | |
Sterile | |
Liver epithelial cells, HaCaT, Primary human hepatocytes, Hybridoma, VERO, HEK293, CHO, HEP G2, HeLa, Human keratinocytes |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.