Learn More
Thermo Scientific™ FastVac™ Vacuum Manifold
The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time.
Brand: Thermo Scientific™ A35899
217.34 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time. Furthermore, the extra-large waste reservoir makes the FastVac Vacuum Manifold convenient for protocols that require the processing of large volumes, such as during plasmid DNA isolation using a PureLink Fast Low-Endotoxin plasmid purification kit.
Product features:
• Capacity—capable of processing 1 to 20 samples simultaneously using columns and a vacuum source
• Composition—consists of a linear, low-density polyethylene manifold body, one-way Luer-Lock stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector, and a neoprene stopper
• Compatibility—compatible with most nucleic acid purification kits (refer to the kit instruction manual for information regarding compatibility and processing with a vacuum manifold)
Specifications
Vacuum Manifold | |
FastVac™ | |
Room Temperature |
• 1 vacuum manifold • 20 Luer-Lock connectors • 1 Neoprene stopper |
|
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.