Learn More
Ohaus™ FD Series Food Portioning Scale
A multi-use portioning scale designed for use in the food service industry
Brand:  Ohaus™ 71170444
 View more versions of this product
Product Code. 11860152
  • 645.00 € / Each
Explore more special offers
Description
Description
The OHAUS FD is a rugged, multi-use portioning scale designed for use in food service applications. Ideally suited for food portioning and checkweighing, the FD combines an easily cleaned design, fast display and high precision load cell. Designed for use in restaurants, bakeries, food manufacturing and other areas requiring fast operation and flexibility, the OHAUS FD scale offers durability and cleanliness in one economical design
  • All stainless steel construction
  • High resolution weighing cells with advanced electronics provide resolution up to 30,000d
  • NSF listed/certified to NSF/ANSI 169-2005 (formerly NSF C-2) and USDA-AMS accepted to NSF/3-A 14159-1, supports HACCP compliance
  • Check Weighing mode with 3 LED indicator and audible alarm
  • Bright backlit LCD display
  • Stable weight display within 2 seconds
  • Multiple weighing units including percent weighing, lb, decimal and fractional oz, kg, g, and lb:oz
  • 120-hour internal rechargeable battery or AC power
  • Industry leading warranty and support
TRUSTED_SUSTAINABILITY
Specifications
Specifications
120 Hr.
User-selectable external span or linearity calibration
Stainless Steel housing and platform, up-front level indicator
5-digit, 7-segment Backlit Blue LCD
4 button membrane switch
FD6
<2 sec.
FD Series Food Portioning Scale
300 mm
71 mm
9 VDC 0.5 A AC Wall Adapter, 100/120 VAC 50/60 Hz
50/60 Hz
Internal rechargeable sealed lead-acid battery
NSF, HACCP
6 kg
0.5 g
±1 g
IP43
-10°C to +40°C
Pound, decimal ounce, 3/4, 1/2 and 1/4 fractional ounce, gram, kilogram, combined lb:oz display (either decimal or fractional ounces)
223 mm
3.6 kg
9/100 to 120 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products