missing translation for 'onlineSavingsMsg'
Learn More

Fehling's reagent II for sugars, For determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline, Honeywell Fluka™

for determination of sugar, Reag. Ph. Eur., solution II: potassium sodium tartrate, alcaline

24.83 € - 2066.48 €

Specifications

CAS 1310-73-2, 304-59-6, 7732-18-5
MDL Number MFCD00012467
UN Number UN1824
View More Specs

Products 10
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15655040
View Documents
Honeywell-Fluka
36019-250ML
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15685040
View Documents
Honeywell-Fluka
36019-500ML
500 mL
62.05 €
500mL
Please to purchase this item. Need a web account? Register with us today!
15635040
View Documents
Honeywell-Fluka
36019-1L
1 L
106.87 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15605050
View Documents
Honeywell-Fluka
36019-6X250ML
6 x 250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15625040
View Documents
Honeywell-Fluka
36019-1.5L
1.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15645040
View Documents
Honeywell-Fluka
36019-2.5L
2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15615050
View Documents
Honeywell-Fluka
36019-6X500ML
6 x 500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15695040
View Documents
Honeywell-Fluka
36019-6X1L
6 x 1 L
589.84 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15675040
View Documents
Honeywell-Fluka
36019-4X2.5L
4 x 2.5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15665040
View Documents
Honeywell-Fluka
36019-30L
30 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

1310-73-2, 304-59-6, 7732-18-5
MFCD00012467
UN1824
Videos
Safety and Handling

Safety and Handling

P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

SDS
Documents

Documents

Product Certifications