missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Felbinac, MedChemExpress
MedChemExpress Felbinac is a potent non-steroidal anti-inflammatory agent, used to treat muscle inflammation and arthritis.
Brand: Medchem Express HY-B0641
This item is not returnable.
View return policy
Description
Felbinac is a potent non-steroidal anti-inflammatory agent, used to treat muscle inflammation and arthritis.Specifications
Felbinac | |
White | |
C14H12O2 | |
4-Biphenylacetic acid | |
O=C(O)CC1=CC=C(C2=CC=CC=C2)C=C1 | |
212.24 | |
Research | |
Neuroscience-Neuromodulation |
5728-52-9 | |
Research | |
1 g | |
DMSO : 100 mg/mL (471.16 mM; Need ultrasonic and warming) | |
212.24 | |
98.07% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction