missing translation for 'onlineSavingsMsg'
Learn More
Fenoldopam hydrochloride, Tocris Bioscience™ Product Code.: 18005222

Fenoldopam hydrochloride, Tocris Bioscience™

Product Code. 18005222
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18005222

Brand: Tocris Bioscience 1659/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective D1-like partial agonist

Selective D1-like dopamine receptor partial agonist (EC50 = 57 nM). Vasodilator in vivo and does not readily cross the blood-brain barrier. Also α2-adrenoceptor antagonist in vitro (Ki = 15 - 25 nM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 181217-39-0
Molecular Formula C16H17Cl2NO3
Molecular Weight (g/mol) 342.216
InChI Key NLMPGIXLXSPNFS-UHFFFAOYSA-N
Synonym fenoldopam hydrochloride, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-1h-3-benzazepine-7,8-diol hydrochloride, skf 82526 hydrochloride, inverted exclamation marka-fenoldopam hydrochloride, inverted exclamation marka-skf 82526 hydrochloride, 6-chloro-1-4-hydroxyphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrochloride, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-4-hydroxyphenyl-, hydrochloride 1:1Show MoreShow Less
PubChem CID 21642479
IUPAC Name 9-chloro-5-(4-hydroxyphenyl)-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol;hydrochloride
SMILES C1CNCC(C2=CC(=C(C(=C21)Cl)O)O)C3=CC=C(C=C3)O.Cl

Specifications

Quantity 10 mg
Formula Weight 342.22
Percent Purity >98%
Chemical Name or Material Fenoldopam hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fenoldopam hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.