missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ FEP Filling Tube
Manufactured from FEP. BRAND™ FEP Filling Tube dispenses accurate volumes directly from drums and bulk refills. For use with Dispensette™III/Organic.
36.35 € - 204.00 €
Product Code | Brand | Overall Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Overall Length (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
10636863
|
BRAND™
704209 |
1.5 m | 6.0 mm |
36.35 €
Each |
In Stock
Log in to see stock. |
||||
10151581
|
BRAND™
704267 |
10 m | 7.6 mm |
204.00 €
Each |
Estimated Shipment: 28-01-2025
Log in to see stock. |
||||
Description
- Manufactured from FEP.
- Dispenses accurate volumes directly from drums and bulk refills.
- For use with Dispensette™III/Organic
Specifications
FEP | |
Dispensette™III/Organic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title