Learn More
Hirschmann™ Discharge Tube Unit with Luer-Lock Connection
Dispensing nozzle with FEP-tubing and Luer-Lock-Connection for use with ceramus™ and opus™ dispenser.
Brand:  Hirschmann™ 9313029
Product Code. 10293621
  • 142.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Guided tubing
  • Screw-coupled
  • With Luer-Lock connector for filtration
Specifications
Specifications
Discharge Tube Unit
FEP, PFA
Ceramus, Opus dos, Battery-Drive ejection unit dispenser
luer-lock connector