Learn More
Hellma FEP Tubing Set
Suitable for use with compact and 3-in-1, flow-through cells with one short and one long screw fitting.
Brand:  Hellma 040-111-722
Product Code. 15776563
  • 84.63 € / 1 set
Estimated Shipment: 08-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fluorinated ethylene propylene (FEP) construction
  • FEP is the preferred material for making small diameter tubing of continuous lengths
  • Chemical and dielectric properties similar to PTFE
  • One short screw fitting
  • One long screw fitting
  • For compact and 3-in-1 cells
Specifications
Specifications
Length: 500 mm
FEP
One short screw connection, one long screw connection
1.1 mm
1.9 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hellma FEP Tubing Set > One short screw connection, one long screw connection