missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5-Fluoroorotic Acid

Detect expression of the URA3 gene, which encodes orotine-5'-monophosphate (OMP) dicarboxylase with Thermo Scientific™ 5-Fluoroorotic Acid, used in yeast molecular genetics.

102.00 € - 417.00 €

Specifications

Chemical Name or Material 5-Fluoroorotic Acid
Product Type 5-Fluoroorotic Acid
Form Powder
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10609920
View Documents
Thermo Scientific™
R0811
1 g
102.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10619920
View Documents
Thermo Scientific™
R0812
5 g
417.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

5-Fluoroorotic Acid is a yeast with an active URA3 gene (Ura+) converts 5-FOA to fluorodeoxyuridine, which is toxic to cells (see the scheme below). Yeast strains carrying a mutation in the URA3 gene grow in the presence of 5-FOA if the media is supplemented with uracil. 5-FOA is frequently used in experiments with Saccharomyces cerevisiae (URA3), Schizosaccharomyces pombe (URA4 and URA5), Candida albicans (URA3), and E. coli (pyrF).

Convertion of 5-FOA to fluorodeoxyuridine

Convertion of 5-FOA to fluorodeoxyuridine

Specifications

Specifications

5-Fluoroorotic Acid
5-Fluoroorotic Acid
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Fluoroorotic acid monohydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 5-Fluoroorotic acidmonohydrate

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.