Learn More
Thermo Scientific™ TAE Buffer (Tris-acetate-EDTA) (50X)
Optimize electrophoresis of both genomic and large supercoiled DNA in agarose and polyacrylamide gels with Thermo Scientific™ 50X TAE Buffer (Tris-acetate-EDTA).
Brand:  Thermo Scientific™ B49
Product Code. 10399519
  • 97.75 € / 1 L
Explore more special offers
Description
Description
Use the Thermo Scientific 50X TAE Buffer (Tris-acetate-EDTA) for electrophoresis of nucleic acids in agarose and polyacrylamide gels. You can use this buffer for both genomic and large supercoiled DNA, and you can also use this as both a running and a gel preparation buffer.
Usage Recommendations
  • Use fresh 1X TAE both for the gel and for the electrophoresis run
  • To prepare 1X TAE buffer add 20ml of 50X TAE buffer to 980ml of deionized water and mix well
Quality Control
  • The absence of endo-, exodeoxyribonucleases and ribonucleases confirmed by appropriate quality tests
1X Composition
  • 40mM Tris
  • 20mM acetic acid
  • 1mM EDTA
  • pHof 50X TAE: 8.4
Recommended for:
Electrophoresis of nucleic acids in agarose and polyacrylamide gels; Used both as a running buffer and as a gel preparation buffer; Filtered through a 0.22μm membrane; Recommended for resolution of RNA and DNA fragments l.
Note:
TAE buffer has a relatively low buffering capacity, therefore periodic replacement of the buffer during prolonged electrophoresis is recommended.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50X
1 L
TAE Buffer
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • TAE Buffer (Tris-acetate-EDTA) (50X)
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetic acid