Learn More
Thermo Scientific™ Buffer Set for Restriction Enzymes
Ensure the optimum reaction conditions for each restriction enzyme with this five buffer system.
Brand:  Thermo Scientific™ B30
Product Code. 11879612
  • 22.31 € / 5mL
Explore more special offers
Description
Description
Our Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. This system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation. The buffer set for restriction endonucleases contains 1 mL of each B, G, O, R, and Tango buffers. It is supplied in 10X concentrated stocks.
Thermo Scientific restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme, while AarI and BveI require an oligonucleotide (also supplied with the enzyme). Esp3I requires DTT.
The following enzymes require unique buffers for optimal digestion: 
  • AarI
  • AjiI
  • BamHI
  • BfuI
  • Bpu10I
  • BseXI
  • Bsp143I
  • Cfr9I
  • Cfr10I
  • Eam1105I
  • Ecl136II
  • Eco52I
  • EcoRI
  • KpnI
  • Lsp1109I
  • PacI
  • PasI
  • Ppu21I
  • SacI
  • ScaI
  • SdaI
  • SduI
  • SgeI
  • TaqI
Tango buffer has been designed for double digestions of DNA with conventional restriction enzymes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5 x 1.0 mL
Restriction Enzyme
Traditional Cloning