missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CpG Methylated Human Genomic DNA

Ensure accurracy in DNA methylation analysis with this methylated human genomic DNA control.

Brand:  Thermo Scientific™ SD1131

Product Code. 11941421

  • 352.00 € / 15µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Human genomic DNA is enzymatically methylated with CpG Methyltransferase (M.SssI). Genomic DNA was isolated from blood samples from healthy male (HIV, HBV, and HCV negative) individuals. Isolated genomic DNA was treated with CpG Methyltransferase, repurified, and dissolved in TE buffer. = 98% CpG methylation level confirmed by bisulfite sequencing.

Highlights:

  • Pure—DNA free from inhibitors, proteins and RNA
  • Convenient—ready to use 100 ng/μL concentration
  • Compatible with downstream applications
  • Complete methylation of cytosines in CpG dinucleotides
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 ng/μL
Sequencing
CpG Methylated Human Genomic DNA
Human
Tube
[other]
15 μg
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2802723 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSD1131
Lot Number2843958 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSD1131
Lot Number2900156 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSD1131
Lot Number2925810 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSD1131
Lot Number2931343 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberSD1131
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ CpG Methylated Human Genomic DNA > 15μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.