Learn More
Thermo Scientific™ DEPC-treated Water
Optimize applications involving RNA with this deionized, diethylpyrocarbonate (DEPC) treated and 0.22μm membrane-filtered water.
Brand:  Thermo Scientific™ R0603
21.28 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
 View more versions of this product
Product Code. 11531575
  • 31.93 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
deionized, diethylpyrocarbonate (DEPC) is treated and 0.22 μm membrane-filtered water. It is ideal for applications involving RNA.
Ideal for applications involving RNA.
  • Free of endo-, exodeoxyribonucleases, ribonucleases and phosphatases
Note:
DEPC-treated Water is not recommended for PCR or in vivo experiments (1).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7.9
Vial
5 x 1 mL
Molecular Biology
DEPC-Treated