missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dNTP/dUTP Mix

Thermo Scientific dNTP/dUTP Mix is a solution containing dATP, dCTP and dGTP, each at a final concentration of 2 mM and dUTP of 4 mM.

Brand:  Thermo Scientific™ R0251

15.76 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10715471

  • 31.51 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific dNTP/dUTP Mix is a solution containing dATP, dCTP and dGTP, each at a final concentration of 2 mM and dUTP of 4 mM. All nucleotides have greater than 99% purity and are free of nuclease activities and human and E. coli DNA. The mix offers the possibility to reduce the number of pipetting steps and the risk of reaction set up errors. dNTP/dUTP Mix is designed for direct use in PCR with Taq DNA Polymerase, reverse transcription, and a wide variety of molecular applications.

Features
• Greater than 99% purity confirmed by HPLC
• Free of human and E. coli DNA

High stability
• Stable for a minimum 100 freeze-thaw cycles
• Up to 95% of dNTPs remain in triphosphate form even after 7 weeks at room temperature
• Up to 90% of dNTPs remain in triphosphate form after 30 cycles of PCR (1 min at 94°C; 3 min at 72°C)

Applications
• cDNA synthesis and RT-PCR
• PCR, qPCR
• Standard PCR
• DNA Sequencing

Note: dNTP/dUTP Mix is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

dNTP/dUTP Mix contains dATP, dCTP, and dGTP at a final concentration of 2 mM and dUTP at 4 mM (1 mL)

Store at –20°C.
DNA Polymerase
>99%
Dry Ice
2 mM
Liquid
Unlabeled
1 mL
Standard PCR, RT-PCR, Real Time PCR (qPCR)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.