missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL)

Thermo Scientific DreamTaq DNA Polymerase is an enhanced Taq DNA polymerase optimized for all standard PCR applications. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase.

Brand:  Thermo Scientific™ EP0702

 View more versions of this product

Product Code. 10160790

  • 103.00 € / 500 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• DreamTaq DNA Polymerase (5 U/μL), 100 μL
• 10X DreamTaq Buffer with 20 mM MgCl2, 2 x 1.25 mL

Store at -20°C.
DreamTaq DNA Polymerase
500 Units
Standard PCR, Genotyping
1X
3'-A
Colorless
Low
Standard
Dry Ice
5 U/μL
No
Separate Components
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Flyers

Reference Materials

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2810511 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0702
Lot Number2814524 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0702
Lot Number2870888 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0702
Lot Number2903733 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0702
Lot Number3037475 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberEP0702
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DreamTaq DNA Polymerase (5 U/μL) > Colorless, 500 Units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.