missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ dUTP Solution (100 mM)

Thermo Scientific dUTP (2'-deoxyuridine 5'-triphosphate) is highly stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Brand:  Thermo Scientific™ R0133

34.33 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 10160780

  • 51.50 € / 0.25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Thermo Scientific dUTP (2'-deoxyuridine 5'-triphosphate) is highly stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. The nucleotide has greater than 99% purity and is free of nuclease activities and human and E. coli DNA. It is designed for many different molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Free of human and E. coli DNA
• Highly stable

Applications
• Long range PCR (40 kb)
• cDNA synthesis and RT-PCR
• Real-time PCR
• Standard PCR
• High fidelity PCR
• Primer extension.

Note: dUTP is not suitable for PCR with proofreading enzymes or enzyme mixes containing proofreading enzymes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

dUTP Solution, 100 mM (0.25 mL)

Store at –20°C.
2-step RT-qPCR, Standard PCR
Unlabeled
0.25 mL
Standard PCR, High-fidelity PCR, RT-PCR, Real Time PCR (qPCR)
Liquid
DNA Polymerase
>99%
Dry Ice
100 mM
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number01248664 Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberR0133
Lot Number2878277 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0133
Lot Number2871043 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0133
Lot Number2890000 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR0133
Lot Number2916057 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberR0133
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ dUTP Solution (100 mM) > 0.25mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.