missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Ecl136II, PacI, SacI Buffer (10X)
Optimize digestions with Ecl136II, PacI, SacI restriction enzymes using this buffer, premixed with BSA for enhanced stability.
Brand: Thermo Scientific™ B26
This item is not returnable.
View return policy
Description
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
restriction enzymes exhibit 100% of their certified activity in the recommended buffer.
The following enzymes require unique buffers for optimal digestion:
- AarI
- AjiI
- BamHI
- BfuI
- Bpu10I
- BseXI
- Bsp143I
- Cfr9I
- Cfr10I
- Eam1105I
- Ecl136II
- Eco52I
- EcoRI
- KpnI
- Lsp1109I
- PacI
- PasI
- Ppu21I
- SacI
- ScaI
- SdaI
- SduI
- SgeI
- TaqI
Specifications
10X | |
1 mL | |
Digestion Buffer | |
Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.