missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Exo-Resistant Random Primer

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of various DNA synthesis reactions.

Brand:  Thermo Scientific™ SO181

58.65 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10334450

  • 77.50 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Exonuclease-Resistant Random Primer is a mixture of single-stranded random oligonucleotides used for highly efficient random priming of various DNA synthesis reactions. This primer has two 3'-terminal phosphorothioate (PTO) modifications that are resistant to the 3'→5' exonuclease activity of proofreading DNA polymerases, such as Klenow Fragment and phi29 DNA Polymerase. It also has 5'- and 3'-hydroxyl ends. The Exo-Resistant Random Primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Features
• 3'-terminal PTO modifications protect from 3'→5' exonuclease degradation and ensure efficient priming of DNA synthesis
• Product delivered in a ready-to-use format

Applications
• Strand displacement amplification of genomic DNA, plasmids, and phage DNA
• DNA labeling by random priming

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Primer
Dry Ice
Random
DNA Labeling, Strand Displacement Amplification
20X Exo-Resistant Random Primer (100 μL)

Store at –20°C.
500 μM
100 μL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Exo-Resistant Random Primer > 100uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.