missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest Bpu10I

Cut at CCTNAGC(-5/-2)^ sites with Thermo Scientific™ FastDigest™ Bpu10l, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer.

Brand:  Thermo Scientific™ FD1184

Product Code. 10202640

  • 86.00 € / 20 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Lambda DNA digested with Bpu10I, 0.7% agarose, 4 cleavage sites

FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment (Cat. No. EP0051), T4 DNA Ligase (Cat. No. EL0014), alkaline phosphatases and T4 DNA Polymerase (Cat. No. EP0061) all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Features
• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available

Applications
• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis

Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.

For methylation sensitivity, refer to product specifications.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• FastDigest Bpu10I (20 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Bpu10I
Yes
No
Restriction Enzyme
FastDigest™
80°C
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
20 μL (20 Reactions)
Traditional Cloning
5' protruding ends
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.