missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluorescein-12-dUTP Solution (1 mM)

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.

Brand:  Thermo Scientific™ R0101

97.50 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10354280

  • 195.00 € / 25µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Fluorescein-12-dUTP (fluorescein-5(6)-carboxaminocaproyl-[5-{3-aminoallyl}-2'-deoxyuridine-5-triphosphate]) is supplied as 1 mM aqueous solution.

Fluorescein-12-dUTP can be enzymatically incorporated into DNA with:
• Reverse transcriptases
Taq DNA polymerase (Cat. No. EP0072)
• phi29 DNA Polymerase (EP0091)
• Klenow Fragment, exo- (EP0421)
• Klenow Fragment (EP0051)
• DNA Polymerase I (EP0041)

Applications
• Enzymatic non-radioactive labeling of DNA during cDNA synthesis
• PCR
• Nick-translation
• Random primed labeling
• Primer extension

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fluorescein-12-dUTP Solution, 1 mM (25 μL)

Store at –20°C.
DNA Polymerase
Fluorescein (FITC)
Dry Ice
1 mM
Liquid
Direct Labeling
25 μL
cDNA Synthesis, Non-Radioactive Labeling
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.