missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant RNA Purification Kit

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

328.00 € - 1336.00 €

Specifications

Isolation Technology Silica Spin Column
Purification Time 20 min. (After sample homogenization)
Elution Volume 50 μL
Sample Type Plants
Final Product Type Total RNA
View More Specs

Requires: Dithiothreitol (DTT, #R0861)


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11591645
View Documents
Thermo Scientific™
K0801
50 Preps
328.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11501655
View Documents
Thermo Scientific™
K0802
250 Preps
1336.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Highlights
Fast—procedure completed in 20 min following cell lysis
Convinient—protocol does not include clogging-prone lysate filtration step
High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)

Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay

Specifications

Specifications

Silica Spin Column
50 μL
Total RNA
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Room Temperature
Up to 65 μg
20 min. (After sample homogenization)
Plants
Not High-throughput Compatible (Manual)
Mini
≤100 mg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneJET Plant RNA Purification Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 C
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine hydrochloride, Thiocyanic acid

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.