missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plasmid Miniprep Kit

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column.

Brand:  Thermo Scientific™ K0503


 View more versions of this product

Product Code. 10242490

  • 365.00 € / 250 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Thermo Scientific GeneJET Plasmid Miniprep Kit utilizes an exclusive silica-based membrane technology in the form of a convenient spin column. The kit recovers up to 20 μg of high copy plasmid DNA per isolation procedure.

Highlights
Efficient—high yields of up to 20 μg of high quality plasmid DNA
Fast—procedure takes less than 14 minutes
Convenient—no phenol/chloroform extraction or alcohol precipitation required
Pure—purified DNA is immediately ready to use

Applications
• Fast isolation of high purity plasmid DNA suitable for all conventional molecular biology procedures, including:
• FastDigest or conventional restriction digestion
• Automated fluorescent and radioactive sequencing
• PCR
In vitro transcription
• Transformation

Principle
A bacterial culture is harvested and lysed. The lysate is then cleared by centrifugation and applied on the silica column to selectively bind DNA molecules at a high salt concentration. The adsorbed DNA is washed to remove contaminants, and the pure plasmid DNA is eluted in a small volume of elution buffer or water. The purified DNA is ready for immediate use in all molecular biology procedures, such as fast and conventional digestion with restriction enzymes, PCR, in vitro transcription, transformation, and automated sequencing. Components Resuspension Solution Lysis Solution Neutralization Solution Wash Solution (concentrated) RNase A Solution Elution Buffer Spin Columns Collection Tubes Detailed Protocol

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spin Column
Silica Column
Bacteria
Compatible (Vacuum manifold)
GeneJET
250 Preps
<14 min
Spin Column
Mini
PCR, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, In Vitro Transcription
< 100 μg (Small-Scale) Plasmid DNA
Plasmid MiniPrep Kit
Plasmid DNA
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium lauryl sulfate, Sodium hydroxide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.