missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Viral DNA/RNA Purification Kit

Perform silica membrane-based viral nucleic acid purification from human and mammalian plasma, serum, blood, and cell-free samples.

Brand:  Thermo Scientific™ K0821


Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11951411

  • 286.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneJET Viral DNA and RNA Purification Kit is designed for rapid and efficient purification of high quality viral nucleic acids from various human and mammalian liquid samples, and is compatible with common sample preservatives like EDTA, citrate, and heparin.

The kit utilizes a silica-based membrane technology in the form of a convenient spin column. Viral nucleic acids from lysed samples bind to the column membrane while impurities are effectively removed during subsequent washing and centrifugation steps. Ready-to-use nucleic acids are eluted from the column.

The purified viral nucleic acids are free of proteins, nucleases, and other contaminants or inhibitors of downstream applications. Isolated DNA and RNA can be directly used in PCR, qPCR, or other nucleic acid based assays.

Highlights:

  • Efficient purification of viral DNA and RNA from human and mammalian plasma, serum, blood, and cell-free samples
  • Linear yields from a wide range of viral titers
  • Tested with a variety of DNA and RNA viruses (HIV, HBV, HCV, HPV, CMV, EBV)
  • Downstream compatibility with (RT)-PCR, (RT)-qPCR, and other nucleic acid based assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spin Column
Silica Spin Column
Milk, Plasma, Saliva, Serum, Swabs (Nasal, Buccal, or Urogenital), Urine, Whole Blood, Virus
Viral DNA and RNA
Not High-throughput Compatible (Manual)
Room Temperature
50 μg (Binding capacity)
• 2 x 1.4 mL Column Preparation Liquid; room temperature
• 12 mL Lysis Solution; room temperature
• 25 mL Wash Buffer 1 (concentrated); room temperature
• 11 mL Wash Buffer 2 (concentrated); room temperature
• 3 x 1.25 mL Eluent; room temperature
• 50 Spin Columns with Wash Tubes; room temperature
• 4 x 50 Wash Tubes; room temperature
• 2 x 1.3 mL Proteinase K; –20°C
• 1 vial Carrier RNA; –20°C

Mini
50 μL
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern/Southern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling, hybridization
50 Preps
30 min.
200 μL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GeneJET Viral DNA/RNA Purification Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Guanidine hydrochloride, Proteinase

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.