Learn More
Thermo Scientific™ GeneJET Whole Blood Genomic DNA Purification Mini Kit
Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation.
171.00 € - 770.00 €
Specifications
Column Type | Spin Column |
---|---|
Scale | Mini |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11521655
|
Thermo Scientific™
K0781 |
50 Reactions |
171.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11531655
|
Thermo Scientific™
K0782 |
250 Preps |
770.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.
Thermo Scientific GeneJET Whole Blood Genomic DNA Purification Mini Kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis and yields purified DNA greater than 30 kb in size. Isolated DNA can be used directly in PCR, Southern blotting, and enzymatic reactions.
Highlights
• Efficient—high yields of high molecular weight genomic DNA
• Fast—20 minute procedure following cell lysis
• Convenient—spin columns are capped and assembled with collection tubes
• Pure—DNA pure from enzyme inhibitors, RNA and proteins. Ready to use for all demanding applications and validated in real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
• Compatible—suitable for wide variety of samples like fresh or frozen anticoagulated mammalian blood (treated with EDTA, Citrate or Heparine), aged blood, buffy coat, bone marrow, and body fluids
Applications
• Isolated genomic DNA is ideal for use in common molecular biology procedures, including:
• PCR and qPCR
• FastDigest and conventional restriction digestion
• Southern blotting Includes Proteinase K Solution Lysis Solution Wash Buffer WB I (concentrated) Wash Buffer II (concentrated) Elution Buffer GeneJET Purification Columns (pre-assembled with collection tubes) Collection tubes Detailed Protocol
Specifications
Spin Column | |
Mini | |
Not High-throughput Compatible (Manual) |
Safety and Handling
- GeneJET Whole Blood Genomic DNA Purification Mini Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Proteinase, Guanidine hydrochloride
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.