Learn More
Thermo Scientific™ GeneRuler 1 kb DNA Ladder
GREENER_CHOICE
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this mixture of chromatography-purified individual DNA fragments.
Brand:  Thermo Scientific™ SM0312
467.40 EUR valid until 2025-12-31
Use promo code "24358" to get your promotional price.
 View more versions of this product
Product Code. 11833963
  • 500.00 € / Pack of 25
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
GeneRuler 1kb DNA Ladder, range 250 to 10,000bp can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye, and it can be used in DNA sizing and approximate quanitification applications.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 1kb DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
6X DNA Loading Dye, 10 x 1mL
25 x 50 μg
Agarose
DNA Ladder
250 to 10000 bp
Sustainable packaging
GeneRuler