missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Genomic DNA Purification Kit

Isolate genomic DNA from various sources, including whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues, with this DNA precipitation system.

Brand:  Thermo Scientific™ K0512

157.70 EUR valid until 2025-12-31
Use promo code "24339" to get your promotional price.



Product Code. 10571031

  • 181.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Genomic DNA Purification Kit is a simple and rapid system for high quality genomic DNA purification from various sources including: whole blood, serum, cell lines, bacterial cells, plant, and mammalian tissues.

The kit is based on selective detergent-mediated DNA precipitation from crude lysate. The entire procedure is rapid only 20 to 25 minutes with a typical yield of 2 to 10 μg genomic DNA from 0.2 mL of blood.

High molecular weight genomic DNA purified with the kit is suitable for direct use in all common molecular biology applications: PCR, FastDigest and conventional restriction digestion, cloning, DNA sequencing, and Southern blot analysis.

Highlights

  • Efficient—2 to 10 μg of genomic DNA from 0.2 mL of blood
  • Fast—procedure takes approximately 20 minutes
  • Universal—purifies genomic DNA from various sources
  • Flexible—easy to scale up and down
  • Safe—does not involve phenol extraction
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 40 mL Lysis Solution; room temperature
• 8 mL Precipitation Solution (concentrated); room temperature
• 10 mL NaCl Solution; room temperature

Bacteria, Cells, Plant, Serum, Tissue, Whole Blood, Blood
gDNA
Not High-throughput Compatible (Manual)
Room Temperature
Bacteria: ≤20 mg
Cells: ≤106
Plant: ≤100 mg
Tissue: ≤30 mg
Serum: ≤200 μL
Whole Blood: ≤200 μL
Organic Extraction
20 to 100 μL
PCR, Southern blotting, sequencing, nucleic acid labeling, hybridization
100 Preps
25 min.
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2812186 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0512
Lot Number2821362 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberK0512
Lot Number2847477 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberK0512
Lot Number2855348 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberK0512
Lot Number2868751 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberK0512
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Genomic DNA Purification Kit > for 100 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.