Learn More
Thermo Scientific™ GTP Solution (100 mM)
Alle ansehen Thermo Scientific ProdukteBeschreibung
Thermo Scientific GTP (guanosine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. It is more than 99% pure and highly stable. GTP may be used in a wide variety of different molecular biology applications.
Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles
Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• in vitro transcription
Spezifikation
Spezifikation
| Concentration | 100 mM |
| For Use With (Application) | in vitro Transcription, RNA Amplification |
| Content And Storage | GTP Solution, 100 mM (0.25 mL) Store at –20°C. |
| Shipping Condition | Dry Ice |
| Conjugate | Unconjugated |
| Quantity | 0.25 mL |
| Product Type | CTP Solution |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.