missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ M13/pUC sequencing primer (-20), 17-mer

Thermo Scientific M13/pUC sequencing primers anneal to the region in the 5'-terminus of the lacZ gene.

Brand:  Thermo Scientific™ SO100

24.24 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10744311

  • 48.47 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific M13/pUC sequencing primers anneal to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 µM aqueous solutions.

Applications
• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19 (Cat. No. SM0221), pTZ19R, pTZ57R, M13mp18, and pBluescript II
• Colony screening by PCR

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sequencing Primer
Dry Ice
M13
Sequencing
M13/pUC sequencing primer (-20), 17-mer, 10 μM

Store at –20°C.
10 μM
pUC19, pTZ19R, pTZ57R, M13mp18, pBluescript II
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.